Learn More
Thermo Scientific™ Pierce™ MMTS (methyl methanethiosulfonate)
Methyl methanethiosulfonate (CAS 2949-92-0); reversibly blocks cysteines (sulfhydryl groups) to facilitate study of enzyme activation and protein functions.
Brand: Thermo Scientific™ 23011
Code : PN
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Pierce MMTS is methyl methanethiosulfonate, a small compound that enables cysteines and other sulfhydryl groups to be reversibly blocked to facilitate study of enzyme activation and other protein functions.
Highlights:
- Converts -SH groups to -S-S-CH3
- Liquid with density of 1.337 (10 M, ∽150 μL)
Specifications
Thiol | |
Unreactive Group | |
Pierce™ | |
Liquid | |
Methyl Methanethiosulfonate |
Upon receipt store at 4°C. | |
Blocking Groups | |
Thiol | |
200 mg |
Safety and Handling
- MMTS (methyl methanethiosulfonate)
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Methanesulfonothioicacid
For Research Use Only. Not for use in diagnostic procedures.