Learn More
Thermo Scientific™ Pierce™ NHS-Activated Agarose Slurry
N-Hydroxysuccinimide ester resin and columns to covalently immobilize proteins, peptides and other amine-containing ligands for affinity purification.
Brand: Thermo Scientific™ 26200
Code : PN
Additional Details : Weight : 0.45360kg
Description
Perform rapid and stable immobilization of proteins, peptides and other ligands with Thermo Scientific™ Pierce NHS-Activated Agarose, a high-quality beaded-agarose support.
The activated affinity resin is available as a slurry or dry powder, allowing simple and efficient immobilization of proteins to a beaded-agarose support. The beaded agarose support contains N-hydroxysuccinimide (NHS) functional groups that react with primary amines to form stable amide linkages. The coupling reaction is performed in an amine-free buffer at pH 7 to 9. Protein coupling efficiency is typically greater than 80%, regardless of the molecular weight or pI the ligand. Coupling capacity exceeds 25mg of protein per milliliter of resin. Once a ligand is immobilized, the prepared resin can be used for multiple affinity purification procedures. The crosslinked beaded agarose has fast linear flow potential, making it useful for gravity-flow and low- to medium-pressure applications.The Pierce NHS-Activated Agarose resin is available in an anhydrous acetone slurry and as dry powder. The unique dry form does not require storage in or removal and disposal of the acetone solvent. In addition, the dry resin is ideal for coupling reactions with dilute samples because it concentrates the sample as the resin swells, reducing the volume of the starting material and resulting in highly efficient ligand immobilization.
Highlights:
Easy to use – immobilize in a simple one-step reaction with minimal hands-on time
Rapid and efficient – greater than 85% coupling for most proteins within 30 minutes
Innovative – the dry agarose concentrates the sample, making it ideal for immobilizing dilute proteins
Safe – no hazardous chemicals needed (e.g., sodium cyanoborohydride, cyanogen bromide)
Versatile – affinity resin is adaptable to column and batch affinity chromatography techniques and FPLC applications
Compatible – use with any primary amine-containing compound
Reusable – the leak-resistant chemistry means you can reuse the affinity resin
High binding capacity – binding capacity of >30mg/mL (slurry) or >25mg/mL (dry resin)
Specifications
≥30 mg per mL Coupling | |
Liquid | |
25 mL | |
Antibody, Protein, Peptide, Ligand | |
NHS Ester | |
Affinity, Agarose Resin |
Upon receipt store the agarose slurry at 4°C. Product is shipped at ambient temperature. | |
Pierce™ | |
Recombinant | |
Agarose | |
Bottle |
Safety and Handling
- Pierce NHS-Activated Agarose Slurry
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetone
For Research Use Only. Not for use in diagnostic procedures.