missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ p-Aminophenyl Phosphoryl Choline Agarose

Product Code. 10753305 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 10753305

Brand: Thermo Scientific™ 20307

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Bind and purify C-reactive protein (CRP) using these agarose beads of immobilized p-aminophenyl phosphoryl choline (APC).

Thermo Scientific™ Pierce p-Amino Phosphoryl Choline Agarose enables the purification of C-reactive protein (CRP).

Immobilized phosphoryl choline is a beaded agarose affinity resin for purification of C-reactive protein (CRP) from plasma, ascites and other biological fluids. CRP is a pentameric (pentraxin) protein that is classified as an acute phase reactant because of its high level of production during inflammation. The protein binds to phosphoryl choline groups on the surface of microbes and has been linked to several biological functions including activation of the classical complement pathway, enhancement of phagocytosis and interaction with certain subpopulations of T-lymphocytes. In the late 1970s, Volanakis and colleagues discovered a method of isolating and studying C-reactive protein by using the protein's affinity for phosphoryl choline that has been immobilized to a porous solid chromatography support.

Highlights:

  • Ligand – p-aminophenyl phosphoryl choline

  • Support – crosslinked 6% beaded agarose

  • Binding capacity – greater than 3mg of human C-reactive protein per milliliter of settled resin

  • Format – 50% resin slurry in 0.02% sodium azide

  • Binding Condition – Tris or borate-buffered saline (pH 8 to 8.5) with 1 to 2mM calcium chloride

  • Elution Condition – Tris or borate-buffered saline (pH 8 to 8.5) with 2mM EDTA

  • Reusable – resin can be regenerated and reused several times

Recommended for:

Purify c-reactive protein (CRP); Purify anti-p-Aminophenyl phosphoryl choline antibodies

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Upon receipt store at 4°C.
Form Liquid
Product Line Pierce
Quantity 5 mL
Target C-reactive Protein
Type Agarose
Stationary Phase p-Amino Phosphoryl Choline
Format Bottle
Column Type Affinity, Agarose Resin
compliance-icons
Product Identifier
  • EZ-Link Micro Sulfo-NHS-Biotinylation Kit
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulfo-NHS-Biotin, Sodium chloride, Sodium phosphate dibasic

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.