Learn More
Thermo Scientific™ Pierce™ Peptide Desalting Spin Columns
Pierce Peptide Desalting Spin Columns provide a convenient and reproducible way to desalt and remove contaminants from peptide samples prior to analysis by mass spectrometry.
Brand: Thermo Scientific™ 89851
342.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific Pierce Peptide Desalting Spin Columns are ready-to-use centrifuge spin columns that enable efficient desalting of peptide samples following enzymatic digestion. The polymer-based hydrophobic resin contained in each column provides excellent binding and recovery characteristics for peptide samples in preparation for mass spectrometry and other methods. Each spin column can bind from 5 μg to 5 mg of native and TMT-labeled peptides. The spin column format allows processing of multiple samples (10–300 μL each) in parallel in approximately 30 minutes.
Mass spectrometry is essential for studying biological compounds. However, many of the buffers and reagents used for sample preparation interfere with mass spectrometry analysis, leading to poor-quality spectra and reduced sensitivity. Pierce Peptide Desalting Spin Columns are designed to remove many of the interfering hydrophilic sample contaminants with minimal sample loss, thus enabling sensitive and comprehensive analysis of digested protein samples by mass spectrometry.• Remove MS-interfering contaminants—significantly reduces signal suppression and improves sensitivity in mass spectrometry
• High binding capacity—hydrophobic polymer-based resin allows for recovery of peptide loads of native and TMT-labeled peptides from 5 μg to 5 mg
• Easy to use—resin is provided in single-use spin column format that fits common 2-mL tubes
• Compatible—resin is validated with a variety of complex samples, including TMT-labeled peptides, and is stable under extreme pH conditions
Specifications
Spin Desalting Column | |
Store at 2–8°C. | |
Peptide | |
Spin Column |
Pierce™ | |
Room Temperature | |
Mass Spectrometry | |
50 Columns |