Learn More
Thermo Scientific™ Pierce™ Peroxidase IHC Detection Kit
Shop All Thermo Scientific ProductsDescripción
Thermo Scientific™ Pierce Peroxidase IHC Detection Kit bundles the Pierce Metal Enhanced DAB Substrate Kit with all necessary components to stain, counterstain and preserve experimental results with frozen or paraffin tissue sections.
Highlights:
Incredible sensitivity – 50 times more sensitive than the traditional DAB method and 30 times more sensitive than other metal-intensified versions of DAB
Low background, high intensity – get a crisp dark brown-black precipitate and, even with the increased intensity, background is almost nonexistent
Six-hour stability – working solution is stable for more than six hours at room temperature! Other DAB substrates must be used immediately
Convenient – eliminates the need to procure numerous reagents from various sources
Includes:
DAB/Metal Concentrate (10X), 25mL; Universal Blocker Blocking Buffer in TBS, 250mL; Peroxidase Suppressor, 2 × 100mL; Stable Peroxide Buffer, 250mL; BupH Tris Buffered Saline, 4 packs; Surfact-Amps 20 (10% Tween-20), 10mL; Harris Modified Hematoxylin, 100mL; Mounting Medium, 60mL
Especificaciones
Especificaciones
| Detection Method | Chromogenic, Colorimetric |
| Quantity | 1 L |
| Content And Storage |
|
| Substrate | DAB (Enhanced) |
| Substrate Type | HRP (Horseradish Peroxidase) Substrate |
| Form | Liquid |
| Product Line | Pierce |
| For Use With (Application) | Immunodetection, IHC (Immunohistochemistry) |
| Product Type | Peroxidase IHC Detection Kit |
For Research Use Only. Not for use in diagnostic procedures.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.