Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor Mini Tablets, EDTA-free
Thermo Scientific™ Pierce Protease Inhibitor Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Brand: Thermo Scientific™ A32955
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Highlights:- Improved formulation&mdsashtablets dissolve quickly into a clear solution
- Economical – more cost-effective than other commercially available formulations for equivalent volumes
- Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
- Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
- EDTA-free- formulated without EDTA, a metalloproteinase inhibitor

Specifications
EDTA-Free | |
1 Vial, Store at 2°C to 8°C | |
Solid | |
10 mL Solution | |
30 Tablets |
Protease Inhibitor | |
Room Temperature | |
Pierce™ | |
Protease Inhibitor |
Safety and Handling
- Pierce Protease Inhibitor Mini Tablets, EDTA-Free
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : Benzenesulfonyl fluoride, 4-(2-aminoethyl)-, hydrochloride
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZI4431702 | Certificate of Analysis | 26/10/2024 | A32955 |
ZC4200413 | Certificate of Analysis | 12/04/2024 | A32955 |
ZA4147961 | Certificate of Analysis | 22/02/2024 | A32955 |
YJ4033512 | Certificate of Analysis | 18/10/2023 | A32955 |
YG3916412 | Certificate of Analysis | 28/07/2023 | A32955 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.