missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ E-64 Protease Inhibitor

Customize your protease inhibitor cocktail formulation with these small packages of individual protease inhibitor peptides and compounds.

Brand:  Thermo Scientific™ 78434

218.83 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


Product Code. 10521004

  • £241.00 / 10mg
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Protease Inhibitors are small packages of individual protease inhibitor peptides and compounds for customized formulation or modification of protease inhibitor cocktails.

The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.

Highlights:

  • High-quality reagents in convenient and affordable package sizes
  • Purchase individually to prepare customized protease inhibitor cocktails
  • Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce E-64 Protease Inhibitor
Store between 2°C and 8°C
Pierce™
Protease Inhibitor
Inhibitor
Lyophilized
Protease Inhibitor
10 mg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.