Learn More
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets,EDTA-free
Thermo Scientific™ Pierce Protease and Phosphatase Inhibitor Mini Tablets broad-spectrum protease and pghospahatase inhibitor formulation provides complete protection from dephosphorylation and proteolytic degradation.
Brand: Thermo Scientific™ A32961
193.12 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Description
The Pierce Protease and Phosphatase Inhibitor Mini Tablets contain aprotinin, bestatin, E-64, EDTA, and leupeptin to provide protection from degradation by serine-proteases, cysteine-proteases, aspartic acid-proteases. To prevent the dephosphorylation activity of serine/threonine and tyrosine phosphatases, each tablet also contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate. Pierce Protease and Phosphatase Inhibitor Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution.
Highlights:- Improved formulation&mdsashtablets dissolve quickly into a clear solution
- Economical – more cost-effective than other commercially available formulations for equivalent volumes
- Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
- Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
- EDTA-free- formulated without EDTA, a metalloproteinase inhibitor
Specifications
Protease and Phosphatase Inhibitor | |
Wet Ice | |
Pierce™ | |
Protease Inhibitor, Phosphatase Inhibitor |
1 Vial, Store at 2°C to 8°C | |
Solid | |
10 mL Solution | |
20 Tablets |
For Research Use Only. Not for use in diagnostic procedures.