missing translation for 'onlineSavingsMsg'
Learn More

Pierce™ Pierce™ Protein A/G Magnetic Agarose Beads

Product Code. 15696144 Shop All Thermo Scientific Products
Click to view available options
Quantity:
4 mL
20 mL
Unit Size:
1mL
5mL
This item is not returnable. View return policy

Product Code. 15696144

Brand: Pierce™ 78610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Pierce™ Protein A/G Magnetic Agarose Beads provide a fast, convenient method for purification of immunoglobulins from serum, cell culture supernatant, or ascites

Product features:

Our recombinant Protein A/G binds to all human IgG subclasses, making it the ideal choice for purification of polyclonal or monoclonal IgG antibodies whose subclass identities have not been determined. In addition, it binds to IgA, IgE, IgM and (to a lesser extent) IgD. Protein A/G binds well to all mouse IgG subclasses but does not bind mouse IgA, IgM, or serum albumin. This makes Protein A/G an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses, without interference from IgA, IgM, and murine serum albumin. Individual subclasses of mouse monoclonals are likely to have a stronger affinity to the chimeric Protein A/G than to either Protein A or Protein G.

  • Protein A/G – immobilized recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables IgG purification from nearly any mammalian species
  • High capacity—sufficient for both routine and demanding purification procedures
  • Low nonspecific binding—optimized purification protocol results in better IgG purification
  • Flexibility—convenience of IgG binding domains of both Protein A and Protein G on one bead
  • Compatibility— beads are compatible with manual and automated applications
  • Inert and stable—superior manufacturing method immobilizes Protein A/G by leach-resistant covalent bonds

Applications:

  • Protein purification and isolation
  • High throughput protein purification using automated magnetic particle processor
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Capacity (Metric) 20 mL
Concentration Slurry: 25%
Content And Storage 5 mL settled resin. Supplied as 20 mL of 25% v/v suspension in phosphate buffered saline, 0.01% Tween-20 detergent, 0.05% sodium azide.

Store at 2–8°C.
Shipping Condition Wet Ice
Particle Size 25 to 30 μm
Form Liquid
Product Line Pierce
Quantity 20 mL
Protein Form Recombinant
Type Magnetic Agarose Bead
Stationary Phase Protein A/G
Format Bottle
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.