Learn More
Thermo Scientific™ Pierce™ Protein L Magnetic Beads
Perform antibody purification using manual or robotic magnetic separators with these high-capacity and high-throughput affinity particles.
Brand: Thermo Scientific™ 88849
Code : PN
Additional Details : Weight : 0.01000kg
Description
Perform antibody purification using manual or robotic magnetic separators with Thermo Scientific™ Pierce Protein L Magnetic Beads, high-capacity and high-throughput affinity particles.
Additional Information and Online Ordering »
Highlights:
Selective – immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
Low non-specific binding – stable, pre-blocked beads provide clean purification of antibody
Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific™ KingFisher Instruments)
Recommended for:
- Purification of monoclonal and polyclonal antibodies that bind poorly to Protein A or Protein G magnetic beads
- Purification of monoclonal antibodies from culture supernatants supplemented with bovine serum
- Protein L IP and co-IP assays
- Purification of ScFv and Fab fragments containing kappa light chains
Specifications
Upon receipt store at 4°C. Product shipped with an ice pack. | |
Pierce™ | |
Recombinant | |
Magnetic Affinity Bead |
Liquid | |
1 mL | |
Antibody | |
Protein L |
For Research Use Only. Not for use in diagnostic procedures.