missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein L Magnetic Beads

Perform antibody purification using manual or robotic magnetic separators with these high-capacity and high-throughput affinity particles.

Brand:  Thermo Scientific™ 88850

 View more versions of this product

Product Code. 13444229

  • £626.00 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce Protein L Magnetic Beads are high-capacity and high-throughput affinity particles for antibody purification using manual or robotic magnetic separators.

Features of Protein L Magnetic Beads:

  • Selective—immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Low non-specific binding—stable, pre-blocked beads provide clean purification of antibody
  • Versatile—beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

Recombinant Protein L, covalently attached to a blocked magnetic bead surface, selectively binds mouse and human antibodies through kappa light chains. The beads are commonly used to purify monoclonal antibodies in cell culture supernatants supplemented with bovine serum as Protein L does not bind bovine IgG. The Pierce Protein L Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms such as the Thermo Scientific KingFisher Instruments for high-throughput workflows.

Applications:

  • Purification of monoclonal and polyclonal antibodies that bind poorly to Protein A or Protein G magnetic beads
  • Purification of monoclonal antibodies from culture supernatants supplemented with bovine serum
  • Protein L IP and co-IP assays
  • Purification of ScFv and Fab fragments containing kappa light chains

Thermo Scientific Pierce Protein L Magnetic Beads are typically used for purifying mouse and human antibodies containing kappa light chains from serum, cell culture supernatant or ascites. Protein L can bind a broader range of Ig classes than Protein A or Protein G, including IgG, IgM, IgA, IgE and IgD. Protein L binds strongly to human (kappa I, III and IV only), mouse (kappa I only), rat and pig immunoglobulins. It binds weakly to rabbit immunoglobulins and does not bind bovine, goat or sheep immunoglobulins. Single chain variable fragments (scFv) and Fab fragments also bind to Protein L. The protocol for the Pierce Protein L beads has been optimized to allow for high recovery and high purity of isolated antibodies.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mL
Upon receipt store at 4°C. Product shipped with an ice pack.
Liquid
5 mL
Antibody
Protein L
Slurry: 10mg/mL, 1% solids
1 μm
Pierce™
Recombinant
Magnetic Affinity Bead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI406198 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number88850
Lot NumberZI397297 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number88850
Lot NumberZC390367 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number88850
Lot NumberZA389976 Certificate TypeCertificate of Analysis Date27/01/2024 Catalog Number88850
Lot NumberSPECS Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number88850
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Protein L Magnetic Beads > 5 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.