missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Quantitative Peroxide Assay Kit (Aqueous

Detect and measure hydrogen peroxide levels (H2O2) in biological samples using an iron (Fe) and xylenol orange (XO) reagent for microplates or tubes.

Brand:  Thermo Scientific™ 23280

217.55 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.



Product Code. 10414195

  • £232.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Sufficient For: 100 tube assays or 500 microplate assays with non-lipid samples
• Reagent A, 1 mL
• Reagent B, 2 x 50 mL

Store at 4°C.
Quantitative Peroxide Assay
Pierce™
Not Target-Specific
Solution-based Detection, Absorbance
100 mL
Pierce Quantitative Peroxide Assay Kit (Aqueous)
Colorimetric
Quantitative Peroxide Assay
100 Tube Assays or 500 Microplate Assays
Spectrophotometer, Microplate Reader
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAA401982 Certificate TypeCertificate of Analysis Date12/02/2025 Catalog Number23280
Lot NumberZJ400573 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number23280
Lot NumberZI388399A Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number23280
Lot NumberZI388399 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number23280
Lot NumberZF392029 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog Number23280
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    55

    Hours

    18

    Minutes

    26

    Seconds

    4
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Quantitative Peroxide Assay Kit (Aqueous > Aqueous-compatible Kit; 100mL kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.