missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rapid ELISA Mouse mAb Isotyping Kit

Isotype mouse monoclonal antibody using an antigen-independent, microplate-based method that enables multiple samples to be tested in about one hour.

Brand:  Thermo Scientific™ 37503

535.80 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.



Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11814524

  • £588.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Sufficient For: 60 isotype determinations (8 wells per test sample)
• Pre-coated 96-well Isotyping Plates, 5 plates
• Goat Anti-Mouse IgG+IgA+IgM-HRP Conjugate, 30 mL
• TMB Substrate, 55 mL
• Stop Solution, 55 mL
• 30X Wash Buffer, 200 mL
• TBS Buffer Packs (each makes 500 mL), 2 packs

Store at 4°C.
Microplate Reader
Isotyping Assay
Pierce™
ELISA (Antigen Capture), Absorbance
60 Tests
Colorimetric
Target-specific
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number429609-003 Certificate TypeCertificate of Analysis Date24/04/2025 Catalog Number
Lot Number429609-002 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number37503
Lot Number429609-001 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number37503
Lot Number390528-009 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number37503
Lot Number390528-008 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog Number37503
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    36

    Hours

    1

    Minutes

    43

    Seconds

    23
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Rapid ELISA Mouse mAb Isotyping Kit > Isotyping Kit; 60-test kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.