Learn More
Thermo Scientific™ Sodium Dodecyl Sulfate (SDS), Lauryl
Perform routine protein electrophoresis and cell lysis methods using this popular, standard-grade SDS detergent.
Brand: Thermo Scientific™ 28365
255.54 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Description
Thermo Scientific™ Sodium Dodecyl Sulfate (Lauryl) is standard-grade SDS detergent for use in protein polyacrylamide gel electrophoresis (PAGE).
Additional Information and Online Ordering
This lauryl-grade sodium dodecyl sulfate (SDS) is a popular anionic detergent for routine protein electrophoresis and cell lysis methods. The formulation is a mixture of several different alkyl sulfate chain lengths (C10 to C18).
Highlights:
- Popular anionic detergent for a variety of protein methods
- Especially useful for denaturing polyacrylamide gel electrophoresis (SDS-PAGE)
- Common component of cell lysis buffers
Detergent properties (pure C12):
- Molecular Weight: 288.5
- Detergent Class: Ionic (anionic)
- Aggregation Number: 62
- Micelle Molecular Weight: 18,000
- Critical Micelle Concentration (CMC): 6 to 8mM (0.1728 to 0.2304%, w/v)
- Cloud Point: greater than 100°C
- Dialyzable: No
Storage Requirements
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
- Should be stored between the following temperatures: 20°C to 25°C
Specifications
White | |
10% (aq, w/v) solution must be clear, colorless to slightly yellow | |
Powder | |
1 kg |
Detergent | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20°C to 25°C. | |
Detergent (Pure) |
Safety and Handling
- Sodium Dodecyl Sulfate (SDS), Lauryl
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium dodecyl sulphate
For Research Use Only. Not for use in diagnostic procedures.