Learn More
Thermo Scientific™ Pierce™ Streptavidin
Purified recombinant streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.
Brand: Thermo Scientific™
325.34 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : PN
Additional Details : CAS Number : 480-65-9 Weight : 0.09000kg
Description
- Purified streptavidin is supplied lyophilized in phosphate-based buffer for immediate reconstitution with water
- Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii
- Pierce™ streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI
- Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin
- Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation
- Applications: ELISA (assay dependent), western blot (assay dependent)
Specifications
Unconjugated | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. | |
Pierce™ | |
Biotin |
5 mg | |
Solid | |
Streptavidin | |
Unlabeled |
For Research Use Only. Not for use in diagnostic procedures.