missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Streptavidin
Purified recombinant streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.
£174.00 - £9780.00
Specifications
Conjugate | Unconjugated |
---|---|
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. |
Product Line | Pierce™ |
Target Molecule | Biotin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10147723
|
Thermo Scientific™
21122 |
1 mg |
£174.00
1mg |
In Stock
Log in to see stock. |
|||||
10700995
|
Thermo Scientific™
21125 |
5 mg |
£514.00
5mg |
In Stock
Log in to see stock. |
|||||
10314654
|
Thermo Scientific™
21135 |
100 mg |
£1348.00
100mg |
In Stock
Log in to see stock. |
|||||
15374870
|
Thermo Scientific™
21145 |
1000 mg |
£9780.00
1000mg |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
Description
- Purified streptavidin is supplied lyophilized in phosphate-based buffer for immediate reconstitution with water
- Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii
- Pierce™ streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI
- Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin
- Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation
- Applications: ELISA (assay dependent), western blot (assay dependent)
Specifications
Unconjugated | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. | |
Pierce™ | |
Biotin |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title