missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin

Purified recombinant streptavidin enables detection of biotinylated antibodies and other probes in a variety of standard assay methods including western blotting, ELISA, immunohistochemistry, and fluorescence imaging.

Brand:  Thermo Scientific™ 21145

8009.88 GBP valid until 2024-12-31
Use promo code "21979" to get your promotional price.


 View more versions of this product

Product Code. 15374870

  • £9230.00 / 1000mg
Estimated Shipment: 03-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Purified streptavidin is supplied lyophilized in phosphate-based buffer for immediate reconstitution with water
  • Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii
  • Pierce™ streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI
  • Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin
  • Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation
  • Applications: ELISA (assay dependent), western blot (assay dependent)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Unconjugated
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Pierce™
Biotin
1000 mg
Powder
Streptavidin Conjugate (fluorescent)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.