Learn More
Thermo Scientific™ Pierce™ Streptavidin Plus UltraLink™ Resin
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Streptavidin Resins comprise a wide selection of high-quality affinity beads and columns for nearly any kind of biotin-based purification procedure.
Additional Information and Online Ordering
Highlights:
Proven coupling chemistries – the methods used to prepare these resins are superior to typical cyanogen bromide (CNBr) immobilization, providing high functionality, low nonspecific binding and excellent leach-resistance
Versatile resins – Agarose (polysaccharide-based) and UltraLink™ Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods
Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)
Superior performance – High Capacity Resin has 2 to 3 times higher binding capacity than alternative supplier products, allowing use of small amounts of resin for experiments
Item-Specific Features:
- Agarose Resins are crosslinked 6% beaded agarose
- UltraLink Resins beaded acrylamide support (UltraLink Biosupport)
- Plus and High Capacity Resins have higher loading (i.e. density) of immobilized streptavidin protein than the regular forms
Recommended for:
Purification of membrane antigens in conjunction with biotinylated monoclonal antibodies; Cell-surface labeling with biotinylation reagents, followed by precipitation with immobilized streptavidin; Purification of cell-surface glycoproteins using biotinylated Concanavalin A; Recovery of single-stranded DNA for dideoxy sequencing
Specifications
Specifications
| Capacity (Metric) | 10 mg Biotinylated BSA Binding per mL Resin |
| Content And Storage | Upon receipt store at 4°C. Product is shipped at ambient temperature. |
| Particle Size | 45 to 165 μm |
| Product Line | Pierce, UltraLink |
| Quantity | 5 mL |
| Protein Form | Recombinant |
| Target | Biotin-Tagged Molecule |
| Type | Resin |
| Stationary Phase | Streptavidin |
| Sufficient For | Binding ≥/= 48 μg Biotin/mL Resin (≥/= 4 mg Biotinylated BSA/mL Resin) |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.