missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Streptavidin UltraLink™ Resin

Bind biotinylated proteins and other ligands with a wide selection of high-quality agarose and acrylamide beads and columns of recombinant streptavidin biotin-binding protein.

£334.00 - £590.00

Specifications

Capacity (Metric) 2 mg Biotinylated BSA Binding per mL Resin
Content And Storage Upon receipt store at 4°C. Product is shipped at ambient temperature.
Particle Size 50 to 80 μm
Product Line Pierce™, UltraLink™
Protein Form Recombinant
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10453644
View Documents
Thermo Scientific™
53113
2 mL
£334.00
2mL
Please to purchase this item. Need a web account? Register with us today!
11879390
View Documents
Thermo Scientific™
53114
5 mL
£590.00
5mL
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

2 mg Biotinylated BSA Binding per mL Resin
50 to 80 μm
Recombinant
Resin
Binding ≥/= 24 μg Biotin/mL Resin (≥/= 2 mg Biotinylated BSA/mL Resin)
Affinity, UltraLink™ Resin
Upon receipt store at 4°C. Product is shipped at ambient temperature.
Pierce™, UltraLink™
Biotin-Tagged Molecule
Streptavidin
Bottle
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Streptavidin UltraLink™ Resin

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.