missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fast Western Kit, SuperSignal™ West Pico, Rabbit

For picogram-level detection and Western blot development less than 1 hour after gel transfer.

Brand:  Thermo Scientific™ 35061

 View more versions of this product

Product Code. 10481945

  • £317.00 / 1 set
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate contains optimized reagents and protocol that allows picogram levels of detection and shortens typical Western blot development to less than 1 hour after the gel transfer.

These Fast Western Blot Kits require minimal hands-on time and yield results comparable to classic Western blotting with SuperSignal West Pico Chemiluminescent Substrate.The kits contain all the reagents necessary to complete a Western blot being probed with either a mouse or rabbit primary antibody. Like other Western blotting methods, the included SuperSignal West Pico Substrate produces a chemiluminescent signal that is detectable using photographic or other imaging methods. The developed blots can be repeatedly exposed and stripped of the primary antibody and reprobed.

Highlights:

  • Fast – picogram levels of detection in about one hour

  • Convenient – no expensive hardware or vacuum required; no clogging issues

  • Simple – optimized protocol makes Western blot analysis easier than ever

  • Economical – no additional consumables or extra equipment to purchase

  • Stable – all kit components are stable for one year stored at 4°C

  • Easy – complete kits contain all components needed to block, probe and develop a blot with any standard mouse or rabbit primary antibody

Compatible with:

Pierce Membranes and Filter Paper; CL-Xposure™ Film for Western blotting; Pierce Western Blot Signal Enhancer; Restore Plus™ Western Blot Stripping Buffer

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce Fast Western Blot Kit, SuperSignal West Pico Substrate, Rabbit
Chemiluminescence
Western Blot
Pierce™, SuperSignal™
HRP
Rabbit
20 Mini Blots
Western Blot Kit
Liquid
Nitrocellulose, PVDF
800 mL
Horseradish Peroxidase
Picogram-level
Sufficient For: 20 mini blots probed with rabbit primary antibodies
• Fast Western Antibody Diluent, 400 mL
• Fast Western 10X Wash Buffer, 200 mL
• Fast Western Rabbit Optimized HRP Reagent, Pico, 20 mL
• SuperSignal West Pico Luminol/Enhancer Solution, 100 mL
• SuperSignal West Stable Peroxide Solution, 100 mL

Store kit at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.