missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade

Product Code. 11511084 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
10 x 1 g
10 x 1 mL
500 mL
Unit Size:
100g
10mL
500mL
Each
Pack of 10
This item is not returnable. View return policy

Product Code. 11511084

Brand: Thermo Scientific™ TS28903

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bottles of 99.5%-pure TFA (MW 114.02, CAS 76-05-01) liquid (sold by weight); for HPLC use as an ion-pairing agent in reverse-phase peptide separations.

Thermo Scientific™ Pierce Trifluoroacetic Acid (TFA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.

For complex peptide separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve peptide that would otherwise overlap. The TFA concentration is usually specified as 0.1% for reverse-phase HPLC of peptides. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make concentrations the same.

Highlights:

  • Purity: >99.5% TFA and exceptional clarity allows sensitive, nondestructive peptide detection at low UV wavelengths
  • High-performance packaging: Packaged under nitrogen in amber glass with protective TFE-lined fluorocarbon caps to ensure TFA integrity
  • Choice of formats for convenience: 1 mL ampules can prepare 1 L of 0.1% v/v TFA solution for the mobile phase in reverse-phase chromatography in moments
  • CAS: 76-05-1

Storage Information

  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
  • Should be stored from 20° to 25°C

Order Info

X in the ordering table indicates that hazardous shipping charges apply.

TRUSTED_SUSTAINABILITY

Specifications

Name Note Trifluoroacetic Acid (TFA)
CAS 76-05-01
Molecular Weight (g/mol) 114.02
Percent Purity 99.5% (TFA)
Quantity 100 g
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic Acid (TFA)
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Trifluoroacetic acid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.