missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Pigment Lance

Pigment lance to sample pigments, pastes and agglomerating bulk goods.

Brand:  Buerkle™ 5369-0310

247.90 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11856403

  • £271.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With half-open flute 50 %, for heavily sticking products
  • With quarter-open flute 25 %, for lightly sticking products
  • Stainless steel AISI 316L (1.4404)
  • 30 mm Ø
Specifications

Specifications

Pigment Lance
Autoclavable
DIN
432 mm
Stainless Steel (V4A, AISI 316)
85 mL
Ideal for Pastes, Pigments and Agglomerating Bulk Goods
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.