Learn More
OHAUS™ Pioneer™ Precision Balance
Affordable balance to achieve reliable results
£1240.00 - £1670.00
Specifications
Connection Type RS232 and USB device (included), GLP/GMP data output with real-time clock
Display Type Backlit LCD
For Use With (Application) Basic weighing, Parts counting, Percent weighing, Animal weighing, Density determination
Material Metal base, Plastic top housing, Removable stainless steel pan, Glass draft shield
Temperature Range (English) 50 to 86 °F, Operability guaranteed between 41 to 104 °F
View More Specs
Products 4
Product Code Brand Capacity Readability Model Price Quantity & Availability  
Product Code Brand Capacity Readability Model Price Quantity & Availability  
17278770
View Documents Promotion Details
OHAUS™
30619385
6200 g 0.01 g PX6202
£1630.00
1 unit
17268770
View Documents
OHAUS™
30619384
6200 g 0.01 g PX6202/E
£1400.00
1 unit
17258770
View Documents
OHAUS™
30619383
620 g 0.001 g PX623
£1670.00
1 unit
17248770
View Documents
OHAUS™
30619382
620 g 0.001 g PX623/E
£1240.00
1 unit
Description
Description
  • Offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point
  • Featuring cast metal lower housing, sub-pan and stainless steel weighing pan
    • Durably constructed for versatile, long-term use
  • Second line display for additional information/guidance and USB/RS232 connectivity for easy communication
    • draft shield models features static removal (ESR) bar for convenient grounding
Key Features:
  • User-selectable environmental filters and brightness settings
  • Auto-tare
  • Auto-dim
  • User-selectable span calibration points
  • Software lockout and reset menu
  • User selectable communication settings and data print options
  • User-definable project and user IDs
Specifications
Specifications
RS232 and USB device (included), GLP/GMP data output with real-time clock
Basic weighing, Parts counting, Percent weighing, Animal weighing, Density determination
50 to 86 °F, Operability guaranteed between 41 to 104 °F
Analytical and precision balances
209 mm
Backlit LCD
Metal base, Plastic top housing, Removable stainless steel pan, Glass draft shield
10 to 30 °C, Operability guaranteed between 5 to 40 °C
321 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
OHAUS™ Pioneer™ Precision Balance