missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Pipemarkers on a Card
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand: Brady™ N007353
This item is not returnable.
View return policy
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Regulations 1271/2008, 790/2009 and 286/2001 | |
Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Gas - Nitrogen | |
Black on Ochre | |
Card | |
Thermal Transfer | |
Nitrogen | |
5 | |
5 Markers/Card | |
-40°F to +194°F | |
0.0049 in. | |
12 x 150 mm | |
150 mm | |
12 mm |
NITROGEN 150X12CARD-T1-P15 | |
Permanent Rubber, Permanent, Pressure-sensitive | |
Warning | |
Polyester | |
Pipe Marker | |
Pipe and valve marking | |
English | |
5 Pk. | |
-40°C to +90°C | |
0.124 mm | |
5.91 in. | |
0.47 in. | |
8 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction