missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Pipemarkers on a Card
European Standard Pipemarkers reinforce safety and aid maintenance.
£11.60
Specifications
Adhesive Type | Permanent Rubber, Permanent, Pressure-sensitive |
---|---|
Format | Card |
Material | Polyester |
Product Type | Pipe Marker |
For Use With (Application) | Pipe and valve marking |
Product Code | Brand | Display Text | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Text | Color | Price | Quantity & Availability | ||||
15359025
|
Brady™
N007344 |
Argon | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15319095
|
Brady™
N007529 |
Carbon dioxide | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15339045
|
Brady™
N007371 |
Bio gas | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15379055
|
Brady™
N007442 |
Condensate | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15309055
|
Brady™
N007433 |
CO2 | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15329105
|
Brady™
N007549 |
Cooling Gas | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15329075
|
Brady™
N007484 |
Freon | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15389085
|
Brady™
N007511 |
Gas | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15389075
|
Brady™
N007493 |
Freon 11 | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15379125
|
Brady™
N007658 |
Helium | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15369105
|
Brady™
N007569 |
Inert Gas | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15329125
|
Brady™
N007625 |
LPG | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15399105
|
Brady™
N007578 |
Mixed Gas | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15329115
|
Brady™
N007587 |
Natural gas | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15359035
|
Brady™
N007353 |
Nitrogen | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15309135
|
Brady™
N007773 |
Return | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
15399115
|
Brady™
N007616 |
Recycled Gas | Black on Ochre |
£11.60
Pack of 5 |
Estimated Shipment: 19-07-2024
Log in to see stock. |
||||
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifications
Permanent Rubber, Permanent, Pressure-sensitive | |
Polyester | |
Pipe and valve marking | |
5 Markers/Card | |
5.91 in. | |
0.47 in. |
Card | |
Pipe Marker | |
5 | |
5 Pk. | |
150 mm | |
12 mm |