Learn More
Brady™ Pipemarkers on a Card
European Standard Pipemarkers reinforce safety and aid maintenance.
Brand:  Brady™ N007578
 View more versions of this product
Product Code. 15399105
  • £12.40 / Pack of 5
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.
Specifications
Specifications
ROHS Environmental Compliance, EU Directive 2002/95/EC, European Regulations 1271/2008, 790/2009 and 286/2001
Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Gas - Mixed gas
Black on Ochre
Card
Thermal Transfer
Mixed Gas
5
5 Markers/Card
-40°F to +194°F
0.0049 in.
12 x 150 mm
150 mm
12 mm
MIXED GAS 150X12CARD-T1-P15
Permanent Rubber, Permanent, Pressure-sensitive
Warning
Polyester
Pipe Marker
Pipe and valve marking
English
5 Pk.
-40°C to +90°C
0.124 mm
5.91 in.
0.47 in.
8 mm