missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperonyl alcohol, 98%, Thermo Scientific Chemicals
£28.80 - £67.00
Chemical Identifiers
CAS | 495-76-1 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.15 |
MDL Number | MFCD00005836 |
InChI Key | BHUIUXNAPJIDOG-UHFFFAOYSA-N |
Synonym | piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol |
PubChem CID | 10322 |
IUPAC Name | 1,3-benzodioxol-5-ylmethanol |
SMILES | C1OC2=C(O1)C=C(C=C2)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10781735
|
Thermo Scientific Acros
131350250 |
25 g | Glass bottle |
£28.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10499440
|
Thermo Scientific Acros
131351000 |
100 g | Glass bottle |
£67.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
495-76-1 | |
152.15 | |
BHUIUXNAPJIDOG-UHFFFAOYSA-N | |
10322 | |
C1OC2=C(O1)C=C(C=C2)CO |
C8H8O3 | |
MFCD00005836 | |
piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol | |
1,3-benzodioxol-5-ylmethanol |
Specifications
495-76-1 | |
White | |
>110°C | |
97.5% min. (GC) | |
MFCD00005836 | |
piperonyl alcohol, piperonol, 1,3-benzodioxole-5-methanol, 3,4-methylenedioxybenzyl alcohol, benzo d 1,3 dioxol-5-ylmethanol, piperonylalcohol, 5-hydroxymethyl-1,3-benzodioxole, 1-hydroxymethyl-3,4-methylenedioxybenzene, benzo 1,3 dioxol-5-ylmethanol, 3,4-methylenedioxy phenylmethanol | |
BHUIUXNAPJIDOG-UHFFFAOYSA-N | |
1,3-benzodioxol-5-ylmethanol | |
10322 | |
98% | |
Piperonyl alcohol |
51.0°C to 55.0°C | |
156.0°C to 158.0°C (66.0 mmHg) | |
Authentic | |
C8H8O3 | |
19, 67 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether, benzene,, soluble in chloroform | |
C1OC2=C(O1)C=C(C=C2)CO | |
152.15 | |
152.15 | |
Crystalline Powder or Crystals |
Safety and Handling
missing translation for 'einecsNumber' : 207-808-4
RUO – Research Use Only