Learn More
Melford PIPES Sodium Salt
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Brand: Melford P40150-500.0
This item is not returnable.
View return policy
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
pKa (25°C): 6.8The working pH range in aqueous solution is 6.5-7.5
Specifications
PIPES, Sodium Salt | |
10010-67-0 | |
500 g | |
C8H17N2NaO6S2 |