missing translation for 'onlineSavingsMsg'
Learn More

Melford PIPES Sodium Salt

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

£133.00 - £3600.00

Specifications

Chemical Name or Material PIPES, Sodium Salt
CAS 10010-67-0
Molecular Formula C8H17N2NaO6S2
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30018699
View Documents
Melford
P40150-100.0
100 g
£133.00
100g
Please to purchase this item. Need a web account? Register with us today!
30018702
View Documents
Melford
P40150-5000.0
5 kg
£3600.00
5kg
Please to purchase this item. Need a web account? Register with us today!
30018700
View Documents
Melford
P40150-1000.0
1 kg
£762.00
1kg
Please to purchase this item. Need a web account? Register with us today!
30018701
View Documents
Melford
P40150-500.0
500 g
£534.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

pKa (25°C): 6.8

The working pH range in aqueous solution is 6.5-7.5

Specifications

Specifications

PIPES, Sodium Salt
10010-67-0
C8H17N2NaO6S2
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.