missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Pipette bulb red, natural rubber, 2 ml
For removing or dispensing small quantities of liquids from Pasteur pipette or droppers.
Brand: DWK Life Sciences 292000205
8.97 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
The combination of the Pasteur pipette and rubber bulb can be used as a teat pipette.
- Easy removal or dispensing of small quantities of liquids from Pasteur pipette or droppers

Specifications
GL 25 | |
White | |
2 mL | |
10/Pk. | |
42.4 mm |
Not Autoclavable | |
Rubber | |
Dispensing liquids; Removing liquids; Adding drops of reagents; Pipetting aid | |
42,4 mm | |
14.3 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Pipette bulb red, natural rubber, 2 ml > 2 mL, Hole diameter 5 mm
Spot an opportunity for improvement?Share a Content Correction