Learn More
Fisherbrand™ Flip & Grip™ Pipette Holder
Hang under a shelf to help maximize your working space. Fisherbrand™ Flip & Grip Pipette Holder accommodates 12 single-channel pipettes or a combination of 3 single and 3 multichannel pipettes.
Brand: Fisherbrand™
Code : 41
Additional Details : Weight : 0.80000kg
Description
- Pipettes can be held in the rubber grips and/or the plunger hooks
- Holds most common brands
- Rubber grips prevent scratching pipette
- Holder rotates for easy pipette selection and storage
- Weighted base
- Fixing screw provided for under shelf option
Specifications
Pipette Holder | |
187 mm |