Learn More
Fisherbrand™ Flip & Grip™ Pipette Holder
Hang under a shelf to help maximize your working space. Fisherbrand™ Flip & Grip Pipette Holder accommodates 12 single-channel pipettes or a combination of 3 single and 3 multichannel pipettes.
£126.00 - £133.00
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
15291588
|
Fisherbrand™
|
Color: Blue |
£128.00
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15380360
|
Fisherbrand™
|
Color: Lime |
£133.00
Each |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||||
15390360
|
Fisherbrand™
|
Color: Magenta |
£126.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
Description
- Pipettes can be held in the rubber grips and/or the plunger hooks
- Holds most common brands
- Rubber grips prevent scratching pipette
- Holder rotates for easy pipette selection and storage
- Weighted base
- Fixing screw provided for under shelf option
Specifications
Pipette Holder | |
187 mm |