Learn More
Fisherbrand™ Flip & Grip™ Pipette Holder
Hang under a shelf to help maximize your working space. Fisherbrand™ Flip & Grip Pipette Holder accommodates 12 single-channel pipettes or a combination of 3 single and 3 multichannel pipettes.
Brand:  Fisherbrand™
Code : 41
Additional Details : Weight : 0.86180kg
 View more versions of this product
Product Code. 15380360
  • £133.00 / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket
Description
Description
  • Pipettes can be held in the rubber grips and/or the plunger hooks
  • Holds most common brands
  • Rubber grips prevent scratching pipette
  • Holder rotates for easy pipette selection and storage
  • Weighted base
  • Fixing screw provided for under shelf option
Specifications
Specifications
Pipette Holder
187 mm