missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Plain Glass Plates with Bonded Spacers

Accessory for use with OmniPAGE mini vertical Electrophoresis systems.

£18.00 - £28.85

Specifications

Type Plain Glass Plate
Gel Dimensions 100 x 100 mm
View More Specs

Products 4
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
11857643
View Documents
Fisherbrand™
VS10PGS0.5
0.5mm bonded spacers
£28.85
Pack of 2
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
11867643
View Documents
Fisherbrand™
VS10PGS0.75
0.75mm bonded spacers
£18.00
Pack of 2
Estimated Shipment: 11-02-2025
to see stock.
Add to basket
 
11887643
View Documents
Fisherbrand™
VS10PGS1.5
1.5mm Bonded Spacers
£18.00
Pack of 2
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
11897643
View Documents
Fisherbrand™
VS10PGS2
2.0mm Bonded Spacers
£18.00
Pack of 2
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
 
Description

Description

  • Accessory for use with OmniPAGE mini vertical Electrophoresis systems
Specifications

Specifications

Plain Glass Plate
100 x 100 mm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.