Learn More
Fisherbrand™ Transparent Covers for S-Line Ultrasonic Cleaning Units
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Transparent Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable plastic.
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 1.00000kg
Description
- Newly developed
- Reduces noise and accelerates the heating-up process
- When turned over, cover can be used as a drip-off basin
- Collected cleaning liquid can be emptied back into the tank
Specifications
PC Board | |
Ultrasonic Bath, S 15 |