Learn More
Fisherbrand™ Transparent Covers for S-Line Ultrasonic Cleaning Units
A practical cover to complete your S-Line Ultrasonic Cleaning Unit. Fisherbrand™ Transparent Covers for S-Line Ultrasonic Cleaning Units are manufactured from durable plastic.
£21.30 - £66.58
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10794323
|
Fisherbrand™
|
Ultrasonic Bath, S 15 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10436212
|
Fisherbrand™
|
Ultrasonic Bath, S 60 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10394022
|
Fisherbrand™
|
Ultrasonic Cleaner, S 300 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10199851
|
Fisherbrand™
|
Ultrasonic Bath, S 30/ S 40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10486782
|
Fisherbrand™
|
Ultrasonic Bath, S 100/S 120 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Newly developed
- Reduces noise and accelerates the heating-up process
- When turned over, cover can be used as a drip-off basin
- Collected cleaning liquid can be emptied back into the tank