Learn More
Kartell™ Plastilab™ Tapered Straight Connectors
For straight connection of tubing with different diameters.
Brand:  Kartell™ 7051200
Code : A0
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 11944349
  • £35.80 / Pack of 10
Estimated Shipment: 19-08-2024
to see stock.
Add to basket
Description
Description
For straight connection of tubings with different diameters.
  • Constructed from polypropylene
  • Autoclavable
Specifications
Specifications
Straight Tapered Tubing Connector
Autoclavable
For straight connection of different diameter tubing
4.5mm bore
8-10 mm
Polypropylene
10/Pk.
Straight