missing translation for 'onlineSavingsMsg'
Learn More
Platinum, 10% on carbon, dry, Thermo Scientific Chemicals Product Code.: 11320064

Platinum, 10% on carbon, dry, Thermo Scientific Chemicals

Product Code. 11320064
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11320064

Brand: Thermo Scientific Alfa Aesar L00469.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum, 10% on carbon, is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones and aromatics. It is also used for hydrogenolysis of cyclopropanes. It is involved in the catalyzes oxidation of alcohols.

Solubility
Insoluble in water.

Notes
Store in a cool location. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

UN Number UN1325
Quantity 1 g
Merck Index 14,7529
Solubility Information Insoluble in water.
Odor Odorless
Chemical Name or Material Platinum, 10% on carbon, dry

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10212151 Certificate TypeCertificate of Analysis Date21/03/2024 Catalog NumberL00469.03,L00469.14,L00469.06
Lot NumberM09437 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00469.03,L00469.14,L00469.06
Lot Number10109755 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL00469.03,L00469.14,L00469.06
Lot NumberM15224 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00469.03,L00469.14,L00469.06
Lot Number10197711 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00469.03,L00469.14,L00469.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum, 10% on carbon, dry, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.