missing translation for 'onlineSavingsMsg'
Learn More
Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals Product Code.: 11389728

Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals

Product Code. 11389728
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11389728

Brand: Thermo Scientific Alfa Aesar 044222.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platinum on Alumina (aluminum oxide) is catalyst material with applications in synthetic chemistry such as hydrogenation of functional groups. catalyst is recommended for selective hydrogenation reactions. The particle size of the catalyst is ideal for allowing fast separation from the reaction mixture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
MDL Number MFCD03458043

Specifications

Appearance Powder
Quantity 100 g
Solubility Information Insoluble in water,moderately soluble in strong mineral acids.
Form Powder
Chemical Name or Material Platinum, 5% on alumina powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX03K034 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog Number044222.06,044222.22,044222.14
Lot NumberP22K050 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog Number044222.06,044222.22,044222.14
Lot NumberY03H002 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044213.22,044222.22,044213.36,044222.14
Lot NumberP28I040 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044222.06,044222.22,044226.18,044222.14
Lot NumberM17I019 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044222.06,044222.22,044226.18,044222.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum, 5% on alumina powder, reduced, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.