missing translation for 'onlineSavingsMsg'
Learn More
Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals Product Code.: 10562601

Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals

Product Code. 10562601
50 g, Glass bottle
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10562601

Brand: Thermo Scientific Acros 195240500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Black
Quantity 50 g
Formula Weight 195.08
Percent Purity 10% (Pt)
Packaging Glass bottle
Form Powder
Chemical Name or Material Platinum on activated carbon

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0451173 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number195240500,195240100
Lot NumberA0389302 Certificate TypeCertificate of Analysis Date07/10/2022 Catalog Number195240010,195240500,195240100
Product Identifier
  • Platinum on activated carbon, 10% Pt, ca. 50% moisture

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum on activated carbon, 10% Pt, ca .50% moisture, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.