missing translation for 'onlineSavingsMsg'
Learn More
Platinum foil, 0.635mm (0.025in) thick, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15455905

Platinum foil, 0.635mm (0.025in) thick, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15455905
25 x 25 mm
Click to view available options
Quantity:
25 x 25 mm
This item is not returnable. View return policy

Product Code. 15455905

Brand: Thermo Scientific Alfa Aesar 000860.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-06-4
Molecular Formula Pt
MDL Number MFCD00011179
Quantity 25 x 25 mm
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES [Pt]
IUPAC Name platinum
Molecular Weight (g/mol) 195.08
PubChem CID 23939
ChEBI CHEBI:33400
Percent Purity 99.9%
Odor Odorless
Weight ≈8.5g/25x25mm
Form Foil
Assay (metals basis)
Chemical Name or Material Platinum foil, 0.635mm (0.025 in.) thick
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ15P20 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000860.FF
Lot NumberK21Q025 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000860.FF
Lot NumberJ18L24 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000860.FF
Lot NumberE12Q22 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000860.FF
Lot NumberH09M14 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number000860.FF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum foil, 0.635mm (0.025in) thick, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.