Learn More
Platinum(II) chloride, Premion™, 99.99+% (metals basis), Pt 73% min, Thermo Scientific Chemicals
£209.04 - £700.49
Chemical Identifiers
CAS | 10025-65-7 |
---|---|
Molecular Formula | Cl2Pt |
Molecular Weight (g/mol) | 265.98 |
MDL Number | MFCD00011181 |
InChI Key | CLSUSRZJUQMOHH-UHFFFAOYSA-L |
IUPAC Name | platinum(2+) dichloride |
SMILES | [Cl-].[Cl-].[Pt++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334748
|
Thermo Scientific Alfa Aesar
043090.03 |
1 g |
£209.04
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344748
|
Thermo Scientific Alfa Aesar
043090.06 |
5 g |
£700.49
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-65-7 | |
265.98 | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
[Cl-].[Cl-].[Pt++] |
Cl2Pt | |
MFCD00011181 | |
platinum(2+) dichloride |
Specifications
581°C (decomposition) | |
Brown-Green | |
Cl2Pt | |
MFCD00011181 | |
14,7528 | |
CLSUSRZJUQMOHH-UHFFFAOYSA-L | |
platinum(2+) dichloride | |
266 | |
Premion™ | |
5.87 g/mL | |
Platinum (II) chloride |
10025-65-7 | |
Powder | |
PtCl2 | |
UN3260 | |
Soluble in ammoniaband hydrochloric acid. Insoluble in water,alcohol and ether. | |
[Cl-].[Cl-].[Pt++] | |
265.98 | |
≥99.99% | |
Pt 73% min | |
(metals basis) |
Safety and Handling
GHS H Statement
H334-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P260-P264b-P272-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H334
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 233-034-1
missing translation for 'rtecsNumber' : TP2275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only