Learn More
Platinum Iridium gauze, 150 mesh woven from 0.043mm (0.0017in) dia wire, Thermo Scientific Chemicals
Filtration, electrodes
£257.28 - £850.23
Specifications
Chemical Name or Material | Platinum Iridium gauze |
---|---|
Name Note | Woven from 0.043mm (0.0017 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Ir; 90:10 wt% |
MDL Number | MFCD02091683 |
Mesh Size | 150 Mesh |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11375237
|
Thermo Scientific Alfa Aesar
040934.FF |
25 x 25 mm |
£257.28
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410351
|
Thermo Scientific Alfa Aesar
040934.FI |
50 x 50 mm |
£850.23
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Filtration, electrodes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Iridium gauze | |
Pt:Ir; 90:10 wt% | |
150 Mesh | |
Wire Cloth |
Woven from 0.043mm (0.0017 in.) dia. wire | |
MFCD02091683 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.