Learn More
Platinum Iridium wire, 1.5mm (0.06in) dia, Thermo Scientific Chemicals
£1127.00 - £4501.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11450101
|
Thermo Scientific Alfa Aesar
044812.BQ |
5 cm |
£1127.00
5cm |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11460101
|
Thermo Scientific Alfa Aesar
044812.BU |
25 cm |
£4501.00
25cm |
Estimated Shipment: 20-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Platinum-iridium wire is used in platinum jewelry, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibers; in metrology and in fabrication of metal microelectrodes. It is used in the electrical stimulation of nervous tissue and in electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.