Learn More
Platinum Iridium wire, 1.5mm (0.06in) dia, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 044812.BQ
1172.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Platinum-iridium wire is used in platinum jewelry, particularly watch cases. They have been used for spinnerets in the manufacture of synthetic fibers; in metrology and in fabrication of metal microelectrodes. It is used in the electrical stimulation of nervous tissue and in electrophysiological recordings. Pt/Ir alloy has been used in mechanical and electrochemical devices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Platinum Iridium wire | |
Pt:Ir; 90:10 wt% | |
5 cm | |
Odorless | |
Wire |
1.5mm (0.06 in.) dia. | |
MFCD02091683 | |
Insoluble in water and acid. | |
≈1.91g/5cm |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
I04Z004 | Certificate of Analysis | 12/11/2021 | 044812.BU,044812.BQ |
D03Y008 | Certificate of Analysis | 12/11/2021 | 044812.BU,044812.BQ |
H03Q19 | Certificate of Analysis | 12/11/2021 | 044812.BU,044812.BQ |
U17D060 | Certificate of Analysis | 12/11/2021 | 044812.BU,044812.BQ |
S07D024 | Certificate of Analysis | 12/11/2021 | 044812.BU,044812.BQ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only