Learn More
Platinum(IV) chloride, 99.9% (metals basis), Pt 57% min, Thermo Scientific Chemicals
Description
Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 370°C (decomposition) |
Color | Red to Brown |
Physical Form | Crystalline |
Quantity | 1 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Soluble in water and acetone. Slightly soluble in ethanol and insoluble in ether. |
Formula Weight | 336.9 |
Sensitivity | Hygroscopic |
Density | 4.303 |
Chemical Name or Material | Platinum(IV) chloride, Pt 57% min. |
Product Suggestions
Customers who viewed this item also viewed
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.