Learn More
Platinum(IV) chloride, 99%, Thermo Scientific Chemicals
£51.76 - £477.38
Chemical Identifiers
CAS | 13454-96-1 |
---|---|
Molecular Formula | Cl4Pt |
Molecular Weight (g/mol) | 336.88 |
MDL Number | MFCD00011182 |
InChI Key | FBEIPJNQGITEBL-UHFFFAOYSA-J |
Synonym | platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4Show More |
PubChem CID | 26031 |
IUPAC Name | tetrachloroplatinum |
SMILES | Cl[Pt](Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10371004
|
Thermo Scientific Acros
195402500 |
250 mg | Glass bottle |
£51.76
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10685832
|
Thermo Scientific Acros
195400010 |
1 g | Glass bottle |
£140.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10541451
|
Thermo Scientific Acros
195400050 |
5 g | Glass bottle |
£477.38
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13454-96-1 | |
336.88 | |
FBEIPJNQGITEBL-UHFFFAOYSA-J | |
26031 | |
Cl[Pt](Cl)(Cl)Cl |
Cl4Pt | |
MFCD00011182 | |
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4Show More | |
tetrachloroplatinum |
Specifications
370.0°C | |
Brown | |
99% | |
PtCl4 | |
platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4 | |
FBEIPJNQGITEBL-UHFFFAOYSA-J | |
tetrachloroplatinum | |
26031 | |
99% |
13454-96-1 | |
Powder | |
Cl4Pt | |
MFCD00011182 | |
Solubility in water: 587g/L (25°C). Other solubilities: alcohol | |
Cl[Pt](Cl)(Cl)Cl | |
336.88 | |
336.89 | |
Platinum(IV) chloride, 99% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 236-645-1
missing translation for 'rtecsNumber' : TP2275500
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.