missing translation for 'onlineSavingsMsg'
Learn More
Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min, Thermo Scientific Chemicals Product Code.: 11301629

Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min, Thermo Scientific Chemicals

Product Code. 11301629
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11301629

Brand: Thermo Scientific Alfa Aesar 012278.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrogenation catalyst. Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groupsPlatinum(IV) oxide monohydrate is used as a catalyst in hydrogenation, hydrogenolysis, dehydrogenation, and oxidation reactions in organic synthesis. It also finds application in ceramic industry, inorganic chemistry and in chemical synthesis. It is utilized for the selective reduction of alkenes without reducing nitro group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Hydrogenation catalyst
  • Suitable for the reduction of double and triple bonds, aromatics, carbonyls, nitriles, and nitro groups
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12137-21-2
Molecular Formula H2O3Pt
Molecular Weight (g/mol) 245.10
MDL Number MFCD00066964
InChI Key SPGAMGILENUIOF-UHFFFAOYSA-N
Synonym dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2oShow MoreShow Less
PubChem CID 11139377
IUPAC Name dioxoplatinum;hydrate
SMILES O.O=[Pt]=O

Specifications

Melting Point 450°C
Color Black to Brown
Physical Form Powder
Quantity 5 g
Linear Formula PtO2·H2O
UN Number UN1479
Merck Index 14,7527
Solubility Information Insoluble in water,acid,aqua regiaSoluble in water and caustic potash solution; Insoluble in alcohol,acid and aqua regia.
Formula Weight 245.10 (227.09 Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 75% min.
Chemical Name or Material Platinum(IV) oxide monohydrate, Adams Catalyst
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN26L027 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog Number012278.03,012278.06
Lot NumberM26I029 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012278.03,012278.06
Lot NumberH10Q17 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012278.03,012278.06
Lot NumberG15P32 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012278.03,012278.06
Lot NumberE14L38 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012278.03,012278.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Platinum(IV) oxide monohydrate, Adams Catalyst, Pt 75% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.