missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum™ SuperFi™ U Multiplex Master Mix

Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use in next-generation sequencing.

Brand:  Invitrogen™ A5140096

 View more versions of this product

Product Code. 17206006

  • £76.25 / 96 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Designed to synthesize multiple custom amplicons for further use in next-generation sequencing
  • The ready-to-use 4X master mix contains uracil-tolerant high-fidelity DNA polymerase with all necessary buffer components and dNTPs for efficient targeted amplification
  • Optimized for high multiplex PCR
  • Enables fast, efficient, and robust amplification
  • Provides high yields
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Transparent
Store at -25°C to -15°C
Platinum™ SuperFi™ U DNA Polymerase
96 Preps
Collibri™
All
4X
gDNA, cDNA
Sars-Covid-2 Sequencing
96 preps
Master Mix
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers