missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Platinum™ SuperFi™ U Multiplex Master Mix
Invitrogen Platinum SuperFi U Multiplex Master Mix is a product designed to synthesize multiple custom amplicons for further use in next-generation sequencing.
£23.75 - £76.25
Specifications
Color | Transparent |
---|---|
Concentration | 4X |
Content And Storage | Store at -25°C to -15°C |
Enzyme | Platinum™ SuperFi™ U DNA Polymerase |
For Use With (Application) | Sars-Covid-2 Sequencing |
Product Code | Brand | No. of Samples | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Samples | Price | Quantity & Availability | |||||
17295996
|
Invitrogen™
A5140024 |
24 preps |
£23.75
24 reactions |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17206006
|
Invitrogen™
A5140096 |
96 preps |
£76.25
96 reactions |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Designed to synthesize multiple custom amplicons for further use in next-generation sequencing
- The ready-to-use 4X master mix contains uracil-tolerant high-fidelity DNA polymerase with all necessary buffer components and dNTPs for efficient targeted amplification
- Optimized for high multiplex PCR
- Enables fast, efficient, and robust amplification
- Provides high yields
Specifications
Transparent | |
Store at -25°C to -15°C | |
Sars-Covid-2 Sequencing | |
Master Mix |
4X | |
Platinum™ SuperFi™ U DNA Polymerase | |
Collibri™ | |
All |